missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine Serum Albumin Monoclonal Antibody (1D11A1)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA550264
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Bovine Serum Albumin Monoclonal Antibody for Western Blot, ELISA
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
Spezifikation
| Bovine Serum Albumin | |
| Monoclonal | |
| 1.94 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P02769 | |
| ALB | |
| Natural Bovine Serum albumin. | |
| 100 μg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG2b |
| ELISA, Western Blot | |
| 1D11A1 | |
| Unconjugated | |
| ALB | |
| ALB; ALB protein; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; Bos d 6; bovine albumin; bovine serum albumin; BSA; cell growth inhibiting protein 42; FDAH; GIG20; GIG42; growth-inhibiting protein 20; MGC179277; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
| Mouse | |
| Protein G | |
| RUO | |
| 280717 | |
| -20°C or -80°C if preferred | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur