Learn More
Tuttnauer Bowie and Dick Test
Alle ansehen Tuttnauer Produkte
Click to view available options
:
Bowie and Dick Test
Beschreibung
- Special program to test steam penetration to porous and hollow loads.
- The cycle includes 3.5 minutes sterilization after pre-vacuum phase.
Spezifikation
Spezifikation
| Produkttyp | Bowie and Dick Test |
| Sterilität | Unsteril |