Learn More
Thermo Scientific Chemicals Bradykinin (1-7)
Beschreibung
Bradykinin Fragment 1-7 is used in research on the specificity and kinetics of kininase(s). Bradykinin dilates blood vessels via the release of prostacyclin, nitric oxide, and Endothelium Derived Hyperpolarizing Factor. Bradykinin is a physiologically and pharmacologically active peptide of the kinin group of proteins, consisting of nine amino acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Reinigungsverfahren | HPLC |
| Inhalt und Lagerung | -30°C to -10°C |
| Format | Pulver |
| Molekulargewicht | 756.85 |
| Produkttyp | Bradykinin (1-7) |
| Menge | 5 mg |
| Kennzeichnung | RUO - research use only |
| Sequenz | Arg-Pro-Pro-Gly-Phe-Ser-Pro |
Nur für Forschungszwecke
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.