missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (BRCA1/1472), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-53177-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for ELISA, Protein Array.
Spezifikation
| BRCA1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| Mouse | |
| 220 kDa | |
| 100 μg | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Phospho Specific, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 672 | |
| Store at 4C. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| BRCA1/1472 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| BRCA1 | |
| Recombinant human BRCA1 protein fragment (aa445-620) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only