missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (RAY) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
223.65 € - 468.30 €
Spezifikation
| Antigen | BRCA1 |
|---|---|
| Klon | RAY |
| Konzentration | 2.76 mg/mL |
| Verdünnung | Western Blot 2-4 ug/ml, Flow Cytometry 1 ug per million cells, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, CyTOF-ready |
| Anwendungen | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18050916
|
Novus Biologicals
NB100-598SS |
0.025 mL |
237.00 € 223.65 € / 25 Mikroliter Sparen 13.35 € 5% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18031584
|
Novus Biologicals
NB100-598 |
0.1 mL |
496.00 € 468.30 € / 0.10 Milliliter Sparen 27.70 € 5% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
BRCA1 Monoclonal specifically detects BRCA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, CyTOF-ready.Spezifikation
| BRCA1 | |
| 2.76 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| P38398 | |
| 672 | |
| Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| RAY | |
| Western Blot 2-4 ug/ml, Flow Cytometry 1 ug per million cells, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| BRCA1 | |
| IgG2a | |
| Protein G purified | |
| 230 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts