missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrdU Staining Kit
Click to view available options
Menge:
50 Objektträger
Packungsgröße:
1 Stück
Beschreibung
Full Product Description
Our BrdU staining system uses a biotinylated monoclonal anti-BrdU, thus eliminating the need for a species-specific secondary antibody. As a result, BrdU staining can be performed in rabbits, rats and mice without problems of crossreactivity or spurious staining due to the detection of endogenous Ig. Streptavidin-peroxidase with DAB is used for signal generation. Nuclei that have incorporated BrdU will stain dark brown.
Reactivity: All species
Applications: Immunohistochemistry (IHC)
Our BrdU staining system uses a biotinylated monoclonal anti-BrdU, thus eliminating the need for a species-specific secondary antibody. As a result, BrdU staining can be performed in rabbits, rats and mice without problems of crossreactivity or spurious staining due to the detection of endogenous Ig. Streptavidin-peroxidase with DAB is used for signal generation. Nuclei that have incorporated BrdU will stain dark brown.
Reactivity: All species
Applications: Immunohistochemistry (IHC)
Spezifikation
Spezifikation
| Beschreibung | BrdU Staining Kit, 50 Slides, Liquid, 6 to 8 pH, Wet Ice Shipping Condition, 2 to 8°C Storage Temperature, DAB Substrate, For Use With Light Microscope, For Cell Analysis, Cellular Imaging, Immunohistochemistry (IHC), IHC Staining and Detection |
| Produkttyp | BrdU Staining Kit |
| Menge | 50 Objektträger |
| Zur Verwendung mit (Anwendung) | Cell Analysis, Cellular Imaging, Immunohistochemistry (IHC), IHC Staining and Detection |
| Nachweisverfahren | Colorimetric |
| Form | Flüssig |
| Spezies | Human |
| Inhalt und Lagerung | 2 °C bis 8 °C |
| Verpackungsmenge | 1 Each |
| Ausreichend für | 50 Slides |
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur