Learn More
BRE Mouse anti-Human, CoraLite 488, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-66356-100UL
Beschreibung
BRE, also named as BRCC45, is a death receptor-associated antiapoptotic protein, which is also involved in DNA-damage repair, and K63-specific deubiquitination. BRE and its XIAP-sustaining mechanism could represent novel targets for anti-cancer therapy. BRE may regulate TNF-alpha signaling through its interactions with TNFRSF1A. It plays a role in homeostasis or cellular differentiation in cells of neural, epithelial and germline origins. BRE gene has four isoforms with MW 42-47 kDa.
Spezifikation
| BRE | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| IgG2a |
| Immunofluorescence | |
| 2E5A5 | |
| CoraLite 488 | |
| 100 μL | |
| 9577 | |
| Liquid |