Learn More
Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
Artikelnummer. 18117417 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
2 Fläschchen
Packungsgröße:
2 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18117417 2 Fläschchen 2 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18117417 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP247117
 Eingeschränkter Artikel
Verfügbar ab: 21-09-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
These lysates have not been subjected to denaturing or reducing conditions. This allows the tissue or cell lysate to be used in a variety of applications; to study protein-protein interaction, ligand binding, ELISA, immunoprecipitation, 1D and 2D gel electrophoresis, and Western blotting for the detection of specific protein targets. For use in 1D and 2D gel electrophoresis, the addition of a denaturing gel loading buffer with reducing agents may be required. Buffer requirements for performing protein-protein interaction and ligand binding studies can vary significantly from RIPA buffer and may require modifications. In most cases, tissue lysates in RIPA buffer can be used, directly in standard ELISA and immunoprecipitation assays. These lysates are proteomic discovery tools. Researchers should validate and optimize for individual use.
The Breast Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Spezifikation
Tissue Breast
Menge 2 Fläschchen
Wirtsspezies Human
Inhalt und Lagerung Store at −80°C. Avoid freeze-thaw cycles.