missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Briakinumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559519
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Briakinumab is a human anti-IL-12 monoclonal antibody being developed for the treatment of a number of T-cell driven autoimmune diseases. It targets and neutralize interleukin-12 and interleukin-23, two proteins associated with inflammation, such as pro-inflammatory interleukins or tumor necrosis factor- alpha. Briakinumab represents a novel approach to treating psoriasis, Multiple Sclerosis, Crohn's Disease and other autoimmune and inflammatory disorders. As of 2011 pharmaceutical development for psoriasis has been discontinued in the U. S. and Europe. Briakinumab targets and neutralizes interleukin-12 and interleukin-23.
Spezifikation
| Briakinumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ABT-874 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur