Beschreibung
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for ELISA
Spezifikation
Spezifikation
| Antigen | BTLA/CD272 |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Klon | BTLA/1528 |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant extracellular human BTLA/CD272 protein (Uniprot: Q7Z69) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |