Learn More
Bupheninhydrochlorid (Diastereomerengemisch), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Buphenine Hydrochloride |
| CAS | 849-55-8 |
| Summenformel | C19 H25 N O2 . Cl H |
| Menge | 500 mg |
| Synonym | Buphenine Hydrochloride, 4-Hydroxy-α-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzenemethanol hydrochloride, Nylidrin hydrochloride, (±)-Nylidrin hydrochloride, Arlidin, Arlidin hydrochloride, Bufedon, Buphedrin, Buphenin hydrochloride, Buphenine hydrochloride, Dilatal, Dilatol, Dilatol hydrochloride, Dilatropon, Dilydrin, NSC 142004, Nylidrine hydrochloride, Opino, Penitardon, Perdilatal, RHC 3432A, Rudilin, Rydrin, SKF 1700A, Suprifen Psb hydrochloride, Tocodilydrin, Tocodrin, Verina, p-Hydroxy-α-[1-[(1-methyl-3-phenylpropyl)amino]ethyl]benzyl alcohol hydrochloride |
| InChI-Formel | InChI=1S/C19H25NO2.ClH/c1-14(8-9-16-6-4-3-5-7-16)20-15(2)19(22)17-10-12-18(21)13-11-17;/h3-7,10-15,19-22H,8-9H2,1-2H3;1H |
| SMILES | Cl.CC(CCc1ccccc1)NC(C)C(O)c2ccc(O)cc2 |
| IUPAC-Name | 4-[1-Hydroxy-2-(4-Phenylbutan-2-ylamino)propyl]phenol;Hydrochlorid |
| Molekulargewicht (g/mol) | 335.87 |
| Formelmasse | 335.1652 |