missing translation for 'onlineSavingsMsg'
Learn More

(±)-Bupivacaine-d9 (butyl-d9), CDN

Product Code. 30320354
Change view
Click to view available options
Menge:
0.005 g
0.01 g
Unit Size:
0.01 Gramm
Product Code. Menge unitSize
30320354 0.01 g 0.01 Gramm
30319644 0.005 g 0.01 Gramm
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30320354

Brand: CDN CDND69330.01G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Bupivacaine N-butyl-d9
CAS 474668-57-0
Summenformel C18 2H9 H19 N2 O
Menge 0.01 g
Synonym 2-Piperidinecarboxamide, 1-(butyl-d9)-N-(2,6-dimethylphenyl)- (9 CI), N-(2,6-Dimethylphenyl)-1-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)piperidine-2-carboxamide, Bupivacaine-d9, Bupivacaine-D9 (N-butyl-D9), 1-(Butyl-d9)-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide
InChI-Formel InChI=1 S/C18H28N2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3/h8-10,16 H,4-7,11-13H2,1-3H3,(H,19,21)/i1D3,4D2,5D2,12D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N1CCCCC1C(=O)Nc2c(C)cccc2C
IUPAC-Name N-(2,6-dimethylphenyl)-1-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)piperidine-2-carboxamide
Molekulargewicht (g/mol) 297.48
Formelmasse 297.2767 g/mol
Reinheit (%) 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.