Learn More
Invitrogen™ Buprenorphine Polyclonal Antibody
GREENER_CHOICE
Sheep Polyclonal Antibody
Marke:  Invitrogen™ PA175056
Artikelnummer. 11554280
  • 794.00 € / 1 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Working Concentration: 0.312 μg/mL. Sensitivity: 10 ng/mL Buprenorphine HCl produces 87% inhibition in a competitive ELISA employing Buprenorphine/Norbuprenorphine polyclonal antibody.
Buprenorphine is a derivative of the opioid alkaloid thebaine that is a more potent and longer lasting analgesic than morphine. It appears to act as a partial agonist at mu and kappa opioid receptors and as an antagonist at delta receptors. The lack of delta-agonist activity has been suggested to account for the observation that buprenorphine tolerance may not develop with chronic use. [National Center for Biotechnology Information. PubChem Compound Database; CID=644073].
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Buprenorphine
Polyclonal
PBS with 0.09% sodium azide; pH 7.1
Sheep
Purified
RUO
Chemical
Antibody
IgG
ELISA
Unconjugated
Buprenex; Buprenorfina; Buprenorphinum; C29H41NO4
Norbuprenorphine (3) CPE.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical