missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BYSL Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15704591 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15704591 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15704591 Lieferant Invitrogen™ Lieferanten-Nr. PA556680

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Immunogen sequence: QQQEELEAEH GTGDKPAAPR ERTTRLGPRM PQDGSDDEDE EWPTLEKAAT MTAAGHHAEV VVDPEDERAI EMFMNKNPPA R Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 83%.

Bystin is expressed as a 2-kb major transcript and a 3.6-kb minor transcript in SNG-M cells and in human trophoblastic teratocarcinoma HT-H cells. Protein binding assays determined that bystin binds directly to trophinin and tastin, and that binding is enhanced when cytokeratins 8 and 18 are present. Immunocytochemistry of HT-H cells showed that bystin colocalizes with trophinin, tastin, and the cytokeratins, suggesting that these molecules form a complex in trophectoderm cells at the time of implantation. Using immunohistochemistry it was determined that trophinin and bystin are found in the placenta from the sixth week of pregnancy. Both proteins were localized in the cytoplasm of the syncytiotrophoblast in the chorionic villi and in endometrial decidual cells at the uteroplacental interface. After week 10, the levels of trophinin, tastin, and bystin decreased and then disappeared from placental villi.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen BYSL
Anwendungen Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gen BYSL
Gen-Zugriffsnummer Q13895
Gen-Alias by the ribosomal protein s6 gene, drosophila, homolog-like; Bys; BYSL; bysl {ECO:0000312; Bystin; bystin like; bystin-like; EMBL:AAH79030.1, ECO:0000312; Enp1; RGD:727959}
Gensymbole BYSL
Wirtsspezies Rabbit
Immunogen Recombinant protein corresponding to Human BYSL. Recombinant protein control fragment (Product #RP-94699).
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 705
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.