Learn More
c-Fos Antibody (2H2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-50037AF700
Artikelnummer. 18731461
  • ONLINE EXCLUSIVE PRICE
    562.00 €
    531.30 € / 0.10 Milliliter
    Sparen 30.70 €
    5% Rabatt
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
c-Fos Monoclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Gene Knock-Out
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
c-Fos
Monoclonal
Alexa Fluor 700
activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos
This c-Fos Antibody (2H2) was developed against full length recombinant human c-Fos protein expressed in and purified from E. coli. [UniProt# P01100]
0.1 mL
Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
2353
Store at 4°C in the dark.
IgG1
Western Blot, Immunohistochemistry, Immunofluorescence, Gene Knock-Out
2H2
50 mM Sodium Borate
Mouse
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen