missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-MAF Monoclonal Antibody (sym0F1), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
191.00 € - 518.00 €
Spezifikation
| Antigen | c-MAF |
|---|---|
| Klon | sym0F1 |
| Konzentration | 5 μL/Test |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
17164103
|
Invitrogen
61-9855-41 |
25 Tests | |||||||
17174103
![]() |
Invitrogen™
61-9855-42 |
100 Tests |
518.00 €
100 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
c-maf is the cellular counterpart of oncogenic v-maf that belongs to the family of basic region leucine zipper domain transcription factors. The leucine-zipper domain is involved in the interaction with LRPICD. There are two forms of human c-maf mRNA, c-maf-long and c-maf-short. It is identified in the genome of the acute transforming avian retrovirus AS42. c-maf targets are IL-4 in Th2 cells, the crystalline genes in lens fiber cells, insulin gene in islet, p53 and L7 where it exerts its transcriptional role through binding to a Maf recognition element (MARE). It regulates Th2 differentiation and lineage-specific hematopoiesis. c-maf is a transcription factor for IL-10 gene expression in LPS-activated macrophages. Chromosomal aberration involving maf is found in some forms of multiple myeloma. It is expressed in myeloma cell lines and resting monocytes/macrophages.Spezifikation
| c-MAF | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| 17132, 4094 | |
| IgG2b κ | |
| Affinity chromatography | |
| MAF |
| sym0F1 | |
| Flow Cytometry | |
| PE-eFluor 610 | |
| Mouse | |
| Human, Mouse | |
| O75444, P54843 | |
| MAF | |
| Primary | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts