missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | c-Myc |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | MYC/7150R |
| Konzentration | 0.2 mg/ml |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Zusammensetzung | 10mM PBS with 0.05% BSA |
| Gen-Alias | avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) |
| Wirtsspezies | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?