missing translation for 'onlineSavingsMsg'
Learn More

C18orf25 Antibody, Novus Biologicals™

Product Code. 18613729 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
100 μg
25 μL
Unit Size:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18613729 25 μL 25 Mikroliter
18655267 100 μg 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613729 Supplier Novus Biologicals Supplier No. NBP26882525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C18orf25 Polyclonal antibody specifically detects C18orf25 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen C18orf25
Anwendungen Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Zusammensetzung PBS (pH 7.2) and 40% Glycerol
Gen-Alias ARKadia-like 1, ARKL1, chromosome 18 open reading frame 25, hypothetical protein LOC147339, MGC12909, MGC87799
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: QDTGATWRTSGLLEELNAEAGHLDPGFLASDKTSGNAPLNEEINIASSDSEVEIVGVQEHARCVHPR
Reinigungsverfahren Protein A purified
Menge 25 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 147339
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.