missing translation for 'onlineSavingsMsg'
Learn More

C3orf80 Antibody, R&D Systems™

Artikelnummer. 18612628 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
25 μL
0.1 mL
Packungsgröße:
0.10 Milliliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18612628 25 μL 25 Mikroliter
18632056 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612628 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP24887225ul

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

C3orf80 Polyclonal antibody specifically detects C3orf80 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3orf80
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Zusammensetzung PBS (pH 7.2), 40% Glycerol
Gen-Alias Chromosome 3 Open Reading Frame 80
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PDDDDDSPALLRDEAAAGSQDSLLDSGG
Reinigungsverfahren Immunogen affinity purified
Menge 25 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 401097
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Produkttyp Antibody
Form Purified
Isotype IgG
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.