missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CDH17. The Cadherin-17 Recombinant Protein Antigen is derived from E. coli. The Cadherin-17 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-88239. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Spezifikation
Spezifikation
| Gene ID (Entrez) | 1015 |
| Spezies | Human |
| Reinigungsverfahren | Chromatography |
| Reinheit | >80% |
| Konzentration | 0.5mg/mL |
| Inhalt und Lagerung | Store at -20°C. Avoid freeze-thaw cycles. |
| Zusammensetzung | PBS and 1M Urea, pH 7.4. |
| Zur Verwendung mit (Anwendung) | Blocking/Neutralizing, Control |
| Gensymbol | CDH17 |
| Markertyp | Unmarkiert |
| Mehr anzeigen |
Nur für Forschungszwecke
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?