missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin A Mouse anti-Human, Clone: CALNA/2353, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-07658-100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Calcineurin A Monoclonal specifically detects Calcineurin A in Human samples. It is validated for Immunohistochemistry-Paraffin.
Spezifikation
| Calcineurin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform | |
| Recombinant full-length human Calcineurin A protein | |
| 100 μg | |
| Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway | |
| 5530 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CALNA/2353 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto