missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kanadische Pestizidmischung 3 100µg/ml in Toluol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemischer Name oder Material | Acequinocyl∣Azadirachtin A∣Azoxystrobin∣Carbaryl∣Clofentezine∣Clothianidin∣Cyantraniliprole∣Dodemorph∣Fluopyram∣Hexythiazox∣Pyrethrins∣Quintozene∣Tebufenozide∣Teflubenzuron |
| CAS | 57960-19-7, 11141-17-6, 131860-33-8, 63-25-2, 74115-24-5, 210880-92-5 |
| Bezeichnung der Analyten oder Bestandteile | Acequinocyl∣Azadirachtin A∣Azoxystrobin∣Carbaryl∣Clofentezine∣Clothianidin∣Cyantraniliprole∣Dodemorph∣Fluopyram∣Hexythiazox∣Pyrethrins∣Quintozene∣Tebufenozide∣Teflubenzuron |
| Versandbedingung | Raumtemperatur |
| Zertifizierung/Konformität | ISO 17034 |
| Solvent or Matrix | Toluol |
| Menge | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acequinocyl 100 μg/mL∣Azadirachtin A 100 μg/mL∣Azoxystrobin 100 μg/mL∣Carbaryl 100 μg/mL∣Clofentezine 100 μg/mL∣Clothianidin 100 μg/mL∣Cyantraniliprole 100 μg/mL∣Dodemorph 100 μg/mL∣Fluopyram 100 μg/mL∣Hexythiazox 100 μg/mL∣Pyrethrins 100 μg/mL∣Quintozene 100 μg/mL∣Tebufenozide 100 μg/mL∣Teflubenzuron 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?