missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Canakinumab Recombinant Monoclonal Antibody
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Canakinumab is a recombinant, human anti-human-IL-1beta monoclonal antibody that belongs to the IgG1/kappa isotype subclass. It is expressed in a murine Sp2/0-Ag14 cell line and comprised of two 447- (or 448-) residue heavy chains and two 214-residue light chains, with a molecular mass of 145157 Daltons when deglycosylated. Both heavy chains of canakinumab contain oligosaccharide chains linked to the protein backbone at asparagine 298 (Asn298). Canakinumab binds to human IL-1beta and neutralizes its inflammatory activity by blocking its interaction with IL-1 receptors, but it does not bind IL-1alpha or IL-1 receptor antagonist (IL-1ra). Canakinumab is marketed under the brand name Ilaris and indicated for patients 4 years of age and older to treat Familial Cold Autoinflammatory Syndrome (FCAS) and Muckle-Wells Syndrome (MWS), which are both part of the Cryopyrin-Associated Periodic Syndromes (CAPS) as well as for patients 2 years of age and older to treat systemic juvenile idiopathic arthritis (SJIA). Clinical trials have established the administration of canakinumab every 2 weeks to be safe and effective, offering a considerable advantage over the existing treatment with the human IL-1 receptor antagonist, anakinra, which must be injected daily and which is often poorly tolerated by patients.
Spezifikation
Spezifikation
| Antigen | Canakinumab |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Gen-Alias | ACZ885 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 1 mg |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?