Learn More
Invitrogen™ Catalase Recombinant Rabbit Monoclonal Antibody (24GB4290)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557354
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Catalase Recombinant Monoclonal Antibody for Western Blot, Flow
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
Spezifikation
| Catalase | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cat | |
| 2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24248, 847 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4290 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04040, P04762 | |
| Cat | |
| Recombinant protein of Mouse Catalase (aa 262-527). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |