missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin E Monoclonal Antibody (186513)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Cathepsin E may have a role in immune function. It is most likely involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation. It may also play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
Specifications
Specifications
| Antigen | Cathepsin E |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 186513 |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with trehalose and no preservative |
| Gen | CTSE |
| Gen-Zugriffsnummer | P70269 |
| Gen-Alias | A430072O03Rik; C920004C08Rik; CATE; Cathepsin E; Cathepsin E form I; Cathepsin E form II; CE; CEA; CEB; CTSE; Ctsea; EC 3.4.23.34; erythrocyte membrane aspartic proteinase; slow-moving proteinase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?