Beschreibung
In acute myeloid leukemia, especially in the M2 subtype, the t(8;21)(q22;q22) translocation is one of the most frequent karyotypic abnormalities. The translocation produces a chimeric gene made up of the 5′-region of the RUNX1 (AML1) gene fused to the 3′-region of the CBFA2T1 (MTG8) gene. The chimeric protein is thought to associate with the nuclear corepressor/histone deacetylase complex to block hematopoietic differentiation. The protein encoded by this gene binds to the AML1-MTG8 complex and may be important in promoting leukemogenesis. Several transcript variants are thought to exist for this gene, but the full-length natures of only three have been described.
Spezifikation
Spezifikation
| Antigen | CBFA2T2 |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konzentration | 0.8 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gen | CBFA2T2 |
| Gen-Zugriffsnummer | O43439, O70374 |
| Gen-Alias | CBFA2T2, EHT, MTG8 like protein, MTG8 related protein 1, MTGR1, p85, Protein CBFA2T2, ZMYND3 |
| Gensymbole | CBFA2T2 |
| Mehr anzeigen |