missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for CyTOF, Flow Cytometry.
Specifications
Specifications
| Antigen | CCR1 |
| Anwendungen | CyTOF, Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 643854R |
| Konjugat | PerCP |
| Verdünnung | Flow Cytometry, CyTOF-ready |
| Zusammensetzung | PBS |
| Gen-Alias | C-C chemokine receptor type 1, CC-CKR-1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R |
| Wirtsspezies | Rat |
| Immunogen | E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession # P51675 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?