missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD-20 Biosimilar Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
477.00 € - 1204.00 €
Spezifikation
| Antigen | CD20 Biosimilar |
|---|---|
| Konzentration | 1 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30283160
![]() |
Invitrogen™
MA559183 |
100 μg |
477.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30283822
|
Invitrogen™
MA559184 |
1 mg |
1204.00 €
1 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term TRU-015.
The CD20 gene, also known as MS4A1, encodes a B cell-specific membrane-bound protein that plays a crucial role in B cell development and function. Located on chromosome 11q12 in humans, the MS4A1 gene spans 16 kb and includes eight exons. CD20 is a part of the membrane-spanning 4-domain family, subfamily A (MS4A), which encompasses 18 tetraspan proteins. The prominent CD20 mRNA variant is 2.8 kb long, although other shorter variants exist. This phosphoprotein, approximately 33-37 kDa, is expressed on the surface of almost all normal and malignant B cells, making it an ideal target for therapeutic antibodies like rituximab. CD20 is known to interact with several proteins such as CD40, MHCII, and CD81, playing a role in B cell receptor signaling and other immune processes. Despite its widespread clinical use in B cell depletion therapies, the precise biological function of CD20 remains partly unresolved.Spezifikation
| CD20 Biosimilar | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts