Beschreibung
Vascular cell adhesion molecule 1 (VCAM1), also known as CD106, is a 110-kDa transmembrane glycoprotein belonging to the immunoglobulin gene superfamily. VCAM1 is expressed by cytokine-activated endothelium, interacts with integrin VLA4 (α4β1) present on the surface of leukocytes, and mediates both adhesion and signal transduction. It is also expressed either constitutively or inducibly in a variety of other cell types, including vascular smooth muscle cells, differentiating skeletal muscle cells, renal and neural epithelial cells, macrophages (Kupffer cells), dendritic cells, and bone marrow stromal cells (PMID: 7507076, 11359843).
Spezifikation
Spezifikation
| Antigen | CD106 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 1.G11B1 |
| Konzentration | 10 μL/Test |
| Konjugat | FITC |
| Wirtsspezies | Mouse |
| Menge | 100 Tests |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 7412 |
| Mehr anzeigen |