Learn More
CD107b Mouse anti-Human, PE, Clone: H4B4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech PE-65053-100TESTS
Beschreibung
The CD107b antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107b. This antibody recognizes human antigen. The CD107b antibody has been validated for the following applications: IF, FC analysis. This antibody is pe.
Spezifikation
| CD107b | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| H4B4 | |
| PE | |
| 100 Tests | |
| 3920 | |
| IgG1 |