missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11b Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 67 publications
268.00 € - 640.00 €
Spezifikation
| Antigen | CD11b |
|---|---|
| Konzentration | 1.00 mg/mL |
| Verdünnung | Western Blot 2 ug/mL. Use reported in scientific literature (PMID 31082627), Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 21422470), Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200. Use reported in multiple pieces of scientific literature (PMID 23980916), Immunohistochemistry-Paraffin 1:400. Use reported in scientific literature (PMID 31022918), Immunohistochemistry-Frozen reported in scientific literature (PMID 23980916), In-situ Hybridization reported in scientific literature (PMID 27133471), Flow (Cell Surface) 1:10 - 1:1000, Single Cell Western 1:10, Dual RNAscope ISH-IHC |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In Situ Hybridization (ISH), Flow Cytometry, In Situ Hybridization (ISH) |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18758753
|
Novus Biologicals
NB110-89474SS |
0.025 mL |
268.00 €
25 Mikroliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
|
18766433
|
Novus Biologicals
NB110-89474 |
0.1 mL |
640.00 €
0.10 Milliliter |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
CD11b Polyclonal specifically detects CD11b in Human, Mouse, Rat, Bovine, C. elegans, Chinese Hamster, Monkey, Rhesus Macaque samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In-situ Hybridization, Single Cell Western, Dual RNAscope ISH-IHC.Spezifikation
| CD11b | |
| Western Blot 2 ug/mL. Use reported in scientific literature (PMID 31082627), Simple Western 1:50, Flow Cytometry reported in scientific literature (PMID 21422470), Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200. Use reported in multiple pieces of scientific literature (PMID 23980916), Immunohistochemistry-Paraffin 1:400. Use reported in scientific literature (PMID 31022918), Immunohistochemistry-Frozen reported in scientific literature (PMID 23980916), In-situ Hybridization reported in scientific literature (PMID 27133471), Flow (Cell Surface) 1:10 - 1:1000, Single Cell Western 1:10, Dual RNAscope ISH-IHC | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Cytoskeleton Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement | |
| ITGAM | |
| IgG | |
| Affinity Purified | |
| Mouse, rat and human. We have feedback that this antibody does not work in human samples with Western blot. Immunogen has 100% homology to bovine and 88% homology with sheep. |
| 1.00 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In Situ Hybridization (ISH), Flow Cytometry, In Situ Hybridization (ISH) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, C. elegans, Chinese Hamster, Monkey, Rhesus Macaque | |
| P11215, P05555 | |
| 3684 | |
| Rabbit Polyclonal CD11b Antibody was made to a synthetic peptide within residues 250-350 of the mouse CD11b protein. [Swiss-Prot# P05555] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 127.2 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts