Learn More
Invitrogen™ CD122/CD132 Biosimilar Recombinant Monoclonal Antibody
Artikelnummer. 30284605
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
30284605 1 mg -
30283665 100 μg 100 Mikrogramm
2 options
Artikelnummer. 30284605
Marke: Invitrogen™ MA558662
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Singapore ASTR patent anti-IL-2R beta/IL-2R gamma .
CD122 and CD132 are integral subunits of the interleukin-2 receptor (IL-2R) complex, which also includes the alpha chain (CD25). They are respectively known as the beta chain (CD122) and the gamma chain (CD132). These genes are expressed in various cell types and are crucial for immunological responses. The heterodimeric CD122/CD132 receptor subunits are vital for the signal transduction of interleukins like IL-2 and IL-15, which are critical for T cell proliferation and activation. CD122, in particular, has an extensive intracellular region, making it indispensable for downstream signaling. Both CD122 and CD132 contribute to the receptor's function by binding IL-2 and other cytokines, facilitating their role in immune regulation and homeostasis. Studies suggest that CD122 and CD132 may be expressed by fibroblast-like synoviocytes (FLS) and dermal fibroblasts (DF), underscoring their broader biological significance beyond immune cells. This receptor complex is also a focal point in cancer immunotherapy strategies due to its role in modulating T regulatory cells (Tregs) and effector T cells.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen CD122/CD132 Biosimilar
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 1 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG1