Learn More
CD134 (OX40) Monoclonal Antibody (ACT35 (ACT-35)), Biotin, eBioscience™, Invitrogen™
Beschreibung
Description: The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50 kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4^+ T cell compartment. Applications Reported: This ACT35 (ACT-35) antibody has been reported for use in flow cytometric analysis. Applications Tested: This ACT35 (ACT-35) antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Spezifikation
Spezifikation
| Antigen | CD134 (OX40) |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | ACT35 (ACT-35) |
| Konzentration | 5 μL/Test |
| Konjugat | Biotin |
| Zusammensetzung | PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 |
| Gen | TNFRSF4 |
| Gen-Zugriffsnummer | P43489 |
| Gensymbole | TNFRSF4 |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.