missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD14 Recombinant Rabbit Monoclonal Antibody (002), PE

Rabbit Recombinant Monoclonal Antibody
99.00 € - 129.00 €
Spezifikation
| Antigen | CD14 |
|---|---|
| Klon | 002 |
| Konzentration | 0.1 mg/mL |
| Inhalt und Lagerung | 4°C, store in dark, DO NOT FREEZE! |
| Anwendungen | Flow Cytometry |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30535444
![]() |
Invitrogen™
MA560230 |
100 Tests |
129.00 €
100 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
30534484
|
Invitrogen™
MA560231 |
25 Tests |
99.00 €
25 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
CD14 Recombinant Monoclonal Antibody for Flow
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.Spezifikation
| CD14 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| RUO | |
| PBS with 0.5% BSA and 0.09% sodium azide | |
| P08571 | |
| 929 | |
| Recombinant Human CD14 protein (Met1-Met344). | |
| Antibody |
| 002 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Cd14 | |
| CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
| Cd14 | |
| Primary | |
| Protein A |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts