Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (MR1), Super Bright™ 600, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody
160.00 € - 391.00 €
Spezifikation
| Antigen | CD154 (CD40 Ligand) |
|---|---|
| Klon | MR1 |
| Konzentration | 0.2 mg/mL |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15963534
|
Invitrogen
63-1541-80 |
25 μg | |||||||
15973534
![]() |
Invitrogen™
63-1541-82 |
100 μg |
391.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Description: The MR1 monoclonal antibody reacts with mouse CD154, a 39 kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle. For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1. Applications Reported: This MR1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR1 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter.
CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.Spezifikation
| CD154 (CD40 Ligand) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P27548 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| MR1 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Armenian Hamster | |
| Mouse | |
| 21947 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.