missing translation for 'onlineSavingsMsg'
Learn More

CD158f Mouse anti-Human, Brilliant Violet 421, Clone: UP-R1, BD

Artikelnummer. 15804959
Ansicht ändern
Click to view available options
Menge:
50 Tests
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15804959 50 Tests 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15804959 Lieferant BD Biosciences Lieferanten-Nr. 566330

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

The UP-R1 monoclonal antibody specifically recognizes CD158f which is also known as KIR2DL5. CD158f is a ñ60 kDa single-pass type I transmembrane glycoprotein that belongs to the KIR (Killer cell Ig-like receptors) family within the Ig gene superfamily. CD158f is encoded by two gene paralogs, KIR2DL5A and KIR2DL5B /that show greater than 99% identity in their coding sequences. /CD158f molecules contain two extracellular C2-type Ig-like domains and a long cytoplasmic domain containing one canonical immunoreceptor tyrosine-based inhibitory motif (ITIM) and another atypical ITIM sequence. Although the exact ligands for CD158f have not yet been identified, the inhibitory function of UP-R1 antibody-crosslinked CD158f was shown to primarily depend upon a Src homology region 2-containing protein tyrosine phosphatase-2 (SHP-2) -dependent pathway. CD158f is highly polymorphic and is expressed on subsets of NK cells and T lymphocytes by some individuals. UP-R1 recognizes cell surface CD158f encoded by the most commonly expressed allele, KIR2DL5A*001 . The UP-R1 antibody has been predicted to recognize CD158f molecules encoded by other transcriptionally-active KIR2DL5 alleles, A*012 and B*00602

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CD158f
Klassifikation Monoclonal
Klon UP-R1
Konjugat Brilliant Violet 421
Zusammensetzung Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gen-Alias CD158F, CD158F1, CD158F2, KI2LA, KIR2DL5, KIR2DL5A, KIR2DL5B
Wirtsspezies Mouse
Immunogen Human CD158f Recombinant Protein
Reinigungsverfahren Affinity Purified
Menge 50 Tests
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.