missing translation for 'onlineSavingsMsg'
Learn More

CD163 Antibody (CL10658), R&D Systems™

Artikelnummer. 18611284 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
100 μg
25 ul
Packungsgröße:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18611284 100 μg 100 Mikroliter
18621955 25 ul 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18611284 Supplier Novus Biologicals Supplier No. NBP321179100ul
 Eingeschränkter Artikel
Verfügbar ab: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Immunohistochemistry∣Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD163
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon CL10658
Konjugat Unconjugated
Verdünnung Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Zusammensetzung PBS, pH 7.2, 40% glycerol
Gen-Alias CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Wirtsspezies Mouse
Immunogen This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of Q86VB7. Actual immunogen sequence is proprietary.
Reinigungsverfahren Protein A purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
Primär oder sekundär Primary
Gen-ID (Entrez) 9332
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Produkttyp Antibody
Form Purified
Isotype IgG1
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.