missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1e Antibody (704407) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals FAB7330P
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD1e Monoclonal antibody specifically detects CD1e in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Spezifikation
| CD1e | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 704407 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD1e molecule, e polypeptide, FLJ17609, membrane-associated, thymocyte antigen CD1E | |
| Mouse myeloma cell line NS0-derived recombinant human CD1e, Met1-Tyr304, Accession # P15812 | |
| 0.1 mL | |
| Immunology | |
| 913 | |
| Store at 4°C in the dark. | |
| IgG1 |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?