Beschreibung
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF, Multiplex Immunoassay
Spezifikation
Spezifikation
| Antigen | CD20 |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | IGEL/773 |
| Konjugat | HRP |
| Zusammensetzung | PBS |
| Gen-Alias | B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant full-length human CD20 protein (Uniprot: P11836 ) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |