Learn More
CD20 Mouse anti-Human, APC, Clone: 2H7, BD
Mouse Monoclonal Antibody
90.95 € - 295.00 €
Spezifikation
| Antigen | CD20 |
|---|---|
| Klon | 2H7 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Konjugat | APC |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15850579
|
BD Biosciences
560900 |
25 Tests |
91.00 € 90.95 € / 1 Stück Sparen 0.05 € 0.06% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15809778
|
BD Biosciences
559776 |
100 Tests |
295.00 €
1 Stück |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Spezifikation
| CD20 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2b κ | |
| Affinity Purified |
| 2H7 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human 6.16c1.3 B cell line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.