Learn More
CD209 Mouse anti-Human, PE, Clone: DCN46, BD
Beschreibung
The DCN46 antibody specifically binds to dendritic cell-specific ICAM-3 grabbing nonintegrin (DC-SIGN or CD209), a type-II membrane protein of approximately 44 kDa with a mannose-binding C-type lectin domain. It is highly expressed on dendritic cells in mucosal tissues. Its sequence is identical to the HIV-1 envelope gp120-binding C-type lectin, and reports suggest that DC-SIGN binds to HIV-1 gp120 and effectively transmits infectious HIV-1 to resting T lymphocytes expressing CD4 and chemokine receptors. The C-type lectin domain of DC-SIGN is also capable of binding other pathogenic viruses, bacteria, and parasites. Reports also suggest that DC-SIGN enables the highly efficient migration of dendritic cells from blood into the tissues. It can interact with ICAM-2, which has a similar sequence as ICAM-3, and is abundantly expressed on vascular and lymphoid endothelium. Thus, DC-SIGN mediates dendritic cells rolling and transendothelial migration, and its interaction with ICAM-2 is essential to specific migratory functions of dendritic cells.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifica
Specifica
| Antigen | CD209 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | DCN46 |
| Konjugat | PE |
| Beschreibung | DC-SIGN |
| Zusammensetzung | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Wirtsspezies | Mouse |
| Immunogen | Human Monocyte Derived DC Cells |
| Reinigungsverfahren | Affinity Purified |
| Vedi altri risultati |
For Research Use Only.
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.