Learn More
CD22 Mouse anti-Human, FITC, Clone: HIB22, BD
Description
The HIB22 monoclonal antibody specifically binds to CD22. CD22 is a 130-140 kDa glycosylated type I integral membrane protein present on the surface of mature B cells. CD22 is expressed in the cytoplasm of virtually all B cells except plasma cells. CD45RO antigen on T cells and CD75 antigen on B cells have been identified as ligands for CD22. CD22 has been reported to participate in B-cell activation and also as an adhesion molecule. Although the immunobiology of this antigen has not been fully elucidated, reports indicate that ligation of CD22 induces constitutive internalization of the molecule followed by complete degradation.
Spécification
Spécification
| Antigen | CD22 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | HIB22 |
| Konjugat | FITC |
| Beschreibung | BL-CAM; Siglec-2; Bgp135; Lyb8; LPAP |
| Zusammensetzung | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Wirtsspezies | Mouse |
| Reinigungsverfahren | Affinity Purified |
| Menge | 25 Tests |
| Afficher plus |
For Research Use Only.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.