Learn More
Invitrogen™ CD24 Monoclonal Antibody (M1/69), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™
Beschreibung
The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24. Applications Reported: This M1/69 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Brilliant Ultraviolet 805 is a tandem dye that emits at 797 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.
Spezifikation
Spezifikation
| Antigen | CD24 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | M1/69 |
| Konzentration | 0.2 mg/mL |
| Konjugat | Brilliant Ultraviolet 805 |
| Zusammensetzung | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gen | Cd24a |
| Gen-Zugriffsnummer | P24807 |
| Gen-Alias | CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.