missing translation for 'onlineSavingsMsg'
Learn More

CD276 (B7-H3) Monoclonal Antibody (M3.2D7), Functional Grade, eBioscience™, Invitrogen™

Artikelnummer. 17121412
Ansicht ändern
Click to view available options
Menge:
25 mg
50 mg
Packungsgröße:
25 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17121412 25 mg 25 Milligramm
17131412 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17121412 Lieferant Affymetrix eBioscience Lieferanten-Nr. 165973025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

The M3.2D7 monoclonal antibody was generated against and reacts with mouse B7-H3, a member of the B7 family of the Ig superfamily proteins. B7-H3 is reported to be expressed by antigen presenting cells and developing bone during embryogenesis. The ligand for B7-H3 has not yet been identified and it is thought that B7-H3 down-regulates T cell functions by engaging an unknown counter receptor on T cells. M3.2D7 stains mouse B7-H3 transfected cells and not mouse spleen cells. M3.2D7 also stains human B7-H3 transfected cells. Applications Reported: The M3.2D7 antibody has been reported for use in flow cytometric analysis. Applications Tested: The M3.2D7 antibody has been tested by flow cytometric analysis of mB7-H3 transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD276 (B7-H3) Monoclonal antibody specifically detects CD276 (B7-H3) in Mouse samples. It is validated for Control, Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen CD276 (B7-H3)
Anwendungen Flow Cytometry, Functional Assay
Klassifikation Monoclonal
Klon M3.2D7
Konzentration 1 mg/mL
Konjugat Functional Grade
Zusammensetzung PBS with no preservative; pH 7.2
Gen CD276
Gen-Zugriffsnummer Q8VE98
Gen-Alias 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7H3; B7-H3; B7RP-2; CD276; CD276 antigen; Cd276 molecule; CD276 molecule tv1; Costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352
Wirtsspezies Rat
Reinigungsverfahren Affinity chromatography
Menge 25 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 102657
Zielspezies Mouse
Inhalt und Lagerung 4° C
Produkttyp Antibody
Form Liquid
Isotype IgG2a
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.