Learn More
CD3 delta Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Artikelnummer. 16836374 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16836374 20 μL 20 Mikroliter
16826374 150 μL 150 Mikroliter
2 options
Artikelnummer. 16836374
Marke: Proteintech 166691AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined.
Spezifikation
Antigen CD3 delta
Anwendungen Immunoprecipitation, Flow Cytometry, Western Blot
Klassifikation Polyclonal
Konzentration 0.13 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen CD3D
Gen-Zugriffsnummer P04234, P04235
Gen-Alias CD3 delta, CD3D, CD3-delta, T cell receptor T3 delta chain, T3D
Gensymbole CD3D
Wirtsspezies Rabbit
Immunogen CD3 delta Fusion Protein Ag10148
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 12500, 915
Zielspezies Mouse, Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG