Learn More
CD3 epsilon Antibody (OKT-3 (muromonab)), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP2-81031PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry.
Spezifikation
| CD3 epsilon | |
| Monoclonal | |
| PE | |
| PBS | |
| CD3E | |
| Human peripheral E+ cells. | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Human | |
| Purified |
| Flow Cytometry | |
| OKT-3 (muromonab) | |
| Flow Cytometry | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Epitope on the epsilon-subunit within the human CD3 complex. | |
| Store at 4°C in the dark. | |
| IgG κ |