Learn More
Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence procedures.
Marke: Epredia™ MS-353-S0
Beschreibung
CD31 is a glycoprotein expressed on endothelial cells and in platelets. It is known to be involved in cell signaling and cell adhesion.Host Species: Mouse
Clone: JC/70A
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat and pig. Others not tested.
Epitope: Not determined
Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia
Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)
Positive Control: Placenta, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunofluorescence
Spezifikation
| CD31 (Endothelial Cell Marker) Ab-1 | |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein corresponding to the extracellular domain downstream of the signal sequence of the CD31 molecule | |
| IVD | |
| Primary | |
| Concentrated |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| JC/70A | |
| Mouse | |
| 100 μL | |
| Oncogenesis | |
| Human | |
| IgG2a κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.