missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD326 Rat anti-Mouse, BB515, Clone: G8.8, BD Horizon™
Rat Monoclonal Antibody
Marke: BD Biosciences 565425
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD326 Monoclonal antibody specifically detects CD326 in Mouse samples. It is validated for Flow cytometry.
Spezifikation
| CD326 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| G8.8 | |
| Brilliant Blue 515 (BB515) | |
| Ep-CAM; EGP; EGP-2; Egp314; GA733-2; TROP1; Tacsd1; Tacstd1; Ly74; gp40 | |
| Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
| IgG2a κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Reseach Use Only.
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur