Beschreibung
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Immunohistochemistry
Spezifikation
Spezifikation
| Antigen | CD34 |
| Anwendungen | Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | CL14579 |
| Konjugat | PE |
| Zusammensetzung | PBS |
| Gen-Alias | CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P28906, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |