Learn More
CD36 Antibody (FA6-152), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Marke: Novus Biologicals NB600-1423
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD36 Monoclonal specifically detects CD36 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
Spezifikation
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Western Blot 1:100-1:2000, Flow Cytometry 2 ug/10^5 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 ug antibody/200 ug protein, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Functional >1 ug/ml, Immunoassay 10 ug/ml | |
| Protein G purified | |
| Purified |
| Monoclonal | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Store at 4°C. | |
| IgG1 |